2-(5-(1-(5-Carboxypentyl)-3,3-dimethylindolin-2-ylidene)penta-1,3-dien-1-yl)-1,3,3-trimethyl-3H-indol-1-ium chloride ≥95%

Supplier: Ambeed
Warning

Synonyms: Cyanine5 carboxylic acid chloride

A403042-5MG A403042-100MG A403042-10MG
77616-742EA 112.27 USD
77616-742 77616-740 77748-809
2-(5-(1-(5-Carboxypentyl)-3,3-dimethylindolin-2-ylidene)penta-1,3-dien-1-yl)-1,3,3-trimethyl-3H-indol-1-ium chloride ≥95%
2-(5-(1-(5-Carboxypentyl)-3,3-dimethylindolin-2-ylidene)penta-1,3-dien-1-yl)-1,3,3-trimethyl-3H-indol-1-ium chloride

Precaución: for Research Use Only

Formula: C₃₂H₃₉ClN₂O₂
MW: 519.11 g/mol
MDL Number: MFCD28142488
CAS Number: 1032678-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR