6-Bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one ≥98%

Supplier: Ambeed
Warning

A402848-1G A402848-5G
77901-000EA 259 USD
77901-000 77900-860
6-Bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one ≥98%
6-Bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one

Precaución: for Research Use Only

Formula: C₈H₅BrFNO₂
MDL Number: MFCD20441932
CAS Number: 355423-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR