(S)-4-Acetoxy-2-aminobutanoic acid hydrochloride ≥95%

Supplier: Ambeed
Warning

Synonyms: H-OAc-Hse-OH·HCl

A402520-100MG A402520-1G A402520-5G
77879-873EA 62 USD
77879-873 77879-874 77879-875
(S)-4-Acetoxy-2-aminobutanoic acid hydrochloride ≥95%
(S)-4-Acetoxy-2-aminobutanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₄
MW: 197.61 g/mol
MDL Number: MFCD07357497
CAS Number: 250736-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR