6-((7-((1-Aminocyclopropyl)methoxy)-6-methoxyquinolin-4-yl)oxy)-N-methyl-1-naphthamide ≥98%

Supplier: Ambeed
Warning

Synonyms: Lucitanib

A410196-10MG A410196-1MG A410196-5MG
77616-784EA 109.18 USD
77616-784 77616-786 77616-788
6-((7-((1-Aminocyclopropyl)methoxy)-6-methoxyquinolin-4-yl)oxy)-N-methyl-1-naphthamide ≥98%
6-((7-((1-Aminocyclopropyl)methoxy)-6-methoxyquinolin-4-yl)oxy)-N-methyl-1-naphthamide

Precaución: for Research Use Only

Formula: C₂₆H₂₅N₃O₄
MW: 443.49 g/mol
MDL Number: MFCD20527751
CAS Number: 1058137-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR