3-(Aminomethyl)oxetan-3-ol ≥98%

Supplier: Ambeed
Danger

A402379-1G A402379-100MG
77572-248EA 156.56 USD
77572-248 77613-224
3-(Aminomethyl)oxetan-3-ol ≥98%
3-(Aminomethyl)oxetan-3-ol

Precaución: for Research Use Only

Formula: C₄H₉NO₂
MW: 103.12 g/mol
MDL Number: MFCD20444451
CAS Number: 1305208-47-2
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR