Methyl indoline-3-carboxylate ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A402218-250MG A402218-100MG
77613-222EA 82.4 USD
77613-222 77684-138
Methyl indoline-3-carboxylate ≥95%, Ambeed.Inc
Methyl indoline-3-carboxylate

Precaución: For research use only.

Formula: C₁₀H₁₁NO₂
MDL Number: MFCD08059272
CAS Number: 39891-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR