6-Bromothiazolo[4,5-c]pyridine ≥95%

Supplier: Ambeed
Danger

A402175-100MG A402175-250MG
77892-829EA 142 USD
77892-829 77892-830
6-Bromothiazolo[4,5-c]pyridine ≥95%
6-Bromothiazolo[4,5-c]pyridine

Precaución: for Research Use Only

Formula: C₆H₃BrN₂S
MDL Number: MFCD22199689
CAS Number: 1234014-66-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR