9-Ethyl-3-nitro-9H-carbazole ≥98%

Supplier: Ambeed
Warning

A409629-25G A409629-5G
77872-648EA 260 USD
77872-648 77872-649
9-Ethyl-3-nitro-9H-carbazole ≥98%
Formula: C₁₄H₁₂N₂O₂
MDL Number: MFCD00022215
CAS Number: 86-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR