N,N-Dimethylformamide diisopropyl acetal ≥95%

Supplier: Ambeed
Danger

Synonyms: N,N,N-Trimethyl-1,1-bis(1-methylethoxy)amine, 1,1-Diisopropoxy-N,N-dimethylmethanamine, 1,1-Diisopropoxytrimethylamine

A401407-100G A401407-1G A401407-5G
77884-050EA 601 USD
77884-050 77884-051 77884-052
N,N-Dimethylformamide diisopropyl acetal ≥95%
N,N-Dimethylformamide diisopropyl acetal

Precaución: for Research Use Only

Formula: C₉H₂₁NO₂
MDL Number: MFCD00008879
CAS Number: 18503-89-4
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR