1,3-Dihydro-2-benzofuran-4-amine ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A401234-250MG A401234-1G
77684-137EA 44 USD
77684-137 77890-825
1,3-Dihydro-2-benzofuran-4-amine ≥98%, Ambeed.Inc
1,3-Dihydro-2-benzofuran-4-amine

Precaución: For research use only.

Formula: C₈H₉NO
MDL Number: MFCD16659158
CAS Number: 98475-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR