Cilastatin Sodium ≥97%

Supplier: Ambeed
Warning

A400757-100MG A400757-1G A400757-250MG
77616-732EA 38.11 USD
77616-732 77616-734 77616-736
Cilastatin Sodium ≥97%
Cilastatin Sodium

Precaución: for Research Use Only

Formula: C₁₆H₂₅N₂O₅S·Na
MDL Number: MFCD08459332
CAS Number: 81129-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR