Genkwanin, 7-methoxyapigenin ≥98%

Supplier: Ambeed
Warning

A400206-10MG A400206-1MG A400206-5MG
77616-726EA 23.13 USD
77616-726 77616-728 77616-730
Genkwanin, 7-methoxyapigenin ≥98%
Genkwanin, 7-methoxyapigenin

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₅
MDL Number: MFCD00017452
CAS Number: 437-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR