2-Bromo-5-fluoro-4-pyridineboronic acid ≥96%

Supplier: Ambeed
Warning

Synonyms: (2-Bromo-5-fluoropyridin-4-yl)boronic acid, 2-Bromo-5-fluoropyridine-4-boronic acid, 2-Bromo-5-fluoro-4-pyridinylboronic acid

A400169-100MG A400169-1G A400169-250MG
77884-044EA 17 USD
77884-044 77884-045 77884-046
2-Bromo-5-fluoro-4-pyridineboronic acid ≥96%
2-Bromo-5-fluoro-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₄BBrFNO₂
MDL Number: MFCD04973353
CAS Number: 1072951-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR