Cy3NS acid ≥97%

Supplier: Ambeed
Warning

Synonyms: Cyanine3 carboxylic acid chloride, 2-(3-(1-(5-Carboxypentyl)-3,3-dimethylindolin-2-ylidene)prop-1-en-1-yl)-1,3,3-trimethyl-3H-indol-1-ium chloride

A400106-1MG A400106-5MG A400106-10MG
77616-722EA 24.72 USD
77616-722 77616-724 77616-720
Cy3NS acid ≥97%
Cy3NS acid

Precaución: for Research Use Only

MW: 456.62 g/mol
MDL Number: MFCD28385499
CAS Number: 1032678-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR