4-Benzylbenzeneboronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

A408970-1G A408970-250MG
77901-823EA 48 USD
77901-823 77901-727
4-Benzylbenzeneboronic acid pinacol ester ≥98%
4-Benzylbenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₃BO₂
MDL Number: MFCD09266180
CAS Number: 911708-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR