3,7-Dimethyloct-6-en-1-yl isobutyrate ≥97%

Supplier: Ambeed
Warning

A408392-5G A408392-100G A408392-25G
77626-822EA 15.45 USD
77626-822 77879-894 77879-895
3,7-Dimethyloct-6-en-1-yl isobutyrate ≥97%
3,7-Dimethyloct-6-en-1-yl isobutyrate

Precaución: for Research Use Only

Formula: C₁₄H₂₆O₂
MW: 226.35 g/mol
MDL Number: MFCD00026443
CAS Number: 97-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR