tert-Butyl 3-(piperidin-3-yl)azetidine-1-carboxylate ≥98%

Supplier: Ambeed
Warning

A408376-1G A408376-100MG A408376-250MG A408376-5G
77594-238EA 164 USD
77594-238 77686-126 77686-127 77686-128
tert-Butyl 3-(piperidin-3-yl)azetidine-1-carboxylate ≥98%
tert-Butyl 3-(piperidin-3-yl)azetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₂
MDL Number: MFCD14581438
CAS Number: 1251006-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR