6-Bromoisoquinoline-1,3(2H,4H)-dione ≥98%

Supplier: Ambeed
Warning

A399223-100MG A399223-1G A399223-250MG
77892-823EA 54 USD
77892-823 77892-824 77892-825
6-Bromoisoquinoline-1,3(2H,4H)-dione ≥98%
6-Bromoisoquinoline-1,3(2H,4H)-dione

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
MDL Number: MFCD12755363
CAS Number: 501130-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR