tert-Butyl 4-(piperidine-4-carbonyl)piperazine-1-carboxylate ≥95%

Supplier: Ambeed
Warning

A408140-100MG A408140-250MG A408140-5G
77672-988EA 36 USD
77672-988 77672-990 77672-992
tert-Butyl 4-(piperidine-4-carbonyl)piperazine-1-carboxylate ≥95%
tert-Butyl 4-(piperidine-4-carbonyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₇N₃O₃
MW: 297.39 g/mol
MDL Number: MFCD06808758
CAS Number: 203520-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR