5-Nitrothiophene-2-carbonyl chloride ≥95%

Supplier: Ambeed
Danger

Synonyms: 5-Nitrothiophene-2-carbonyl Chloride

A407890-1G A407890-250MG
77686-124EA 57 USD
77686-124 77686-125
5-Nitrothiophene-2-carbonyl chloride ≥95%
5-Nitrothiophene-2-carbonyl chloride

Precaución: for Research Use Only

Formula: C₅H₂ClNO₃S
MW: 191.59 g/mol
MDL Number: MFCD09260988
CAS Number: 39978-57-9
UN: 2928
ADR: 6.1,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR