7-Mercapto-4-methylcoumarin ≥98%

Supplier: Ambeed
Warning

A407852-250MG A407852-1G
77626-820EA 57.68 USD
77626-820 77878-154
7-Mercapto-4-methylcoumarin ≥98%
7-Mercapto-4-methylcoumarin

Precaución: for Research Use Only

Formula: C₁₀H₈O₂S
MDL Number: MFCD00467591
CAS Number: 137215-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR