(2,4-Difluoro-6-methoxyphenyl)boronic acid ≥97%

Supplier: Ambeed
Warning

A398576-1G A398576-100MG A398576-250MG
77572-232EA 160.68 USD
77572-232 77616-718 77746-800
(2,4-Difluoro-6-methoxyphenyl)boronic acid ≥97%
(2,4-Difluoro-6-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₃
MW: 187.94 g/mol
MDL Number: MFCD22414936
CAS Number: 1808997-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR