5-Chloro-2-hydroxybenzyl alcohol ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-2-(hydroxymethyl)phenol

A407614-5G A407614-25G
77594-236EA 44.08 USD
77594-236 77746-822
5-Chloro-2-hydroxybenzyl alcohol ≥98%
5-Chloro-2-hydroxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₇ClO₂
MW: 158.59 g/mol
MDL Number: MFCD00021967
CAS Number: 5330-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR