3-(3-Methyl-3H-diaziren-3-yl)propan-1-ol ≥95%

Supplier: Ambeed
Danger

Synonyms: 3-(3-Methyl-3H-diazirin-3-yl)propan-1-ol

A398203-100MG A398203-50MG
77613-208EA 337.84 USD
77613-208 77686-104
3-(3-Methyl-3H-diaziren-3-yl)propan-1-ol ≥95%
3-(3-Methyl-3H-diaziren-3-yl)propan-1-ol

Precaución: for Research Use Only

Formula: C₅H₁₀N₂O
MW: 114.146 g/mol
MDL Number: MFCD27995612
CAS Number: 16297-94-2
UN: 2924
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR