Methyl 2-(benzyl-methyl-amino)ethyl2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate ≥98%

Supplier: Ambeed
Warning

Synonyms: Nicardipine

A407345-1G A407345-250MG
77601-790EA 216.28 USD
77601-790 77668-854
Methyl 2-(benzyl-methyl-amino)ethyl2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate ≥98%
Methyl 2-(benzyl-methyl-amino)ethyl2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate

Precaución: for Research Use Only

Formula: C₂₆H₂₉N₃O₆
MW: 479.52 g/mol
MDL Number: MFCD00216027
CAS Number: 55985-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR