1,2,4-Tribromobenzene ≥98%

Supplier: Ambeed
Warning

A407323-1G A407323-25G A407323-5G
77886-088EA 7 USD
77886-088 77886-089 77886-090
1,2,4-Tribromobenzene ≥98%
1,2,4-Tribromobenzene

Precaución: for Research Use Only

Formula: C₆H₃Br₃
MDL Number: MFCD00000062
CAS Number: 615-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR