4,4'-(Propane-2,2-diyl)bis(2-isopropylphenol) ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A398121-1G A398121-250MG
77684-132EA 90 USD
77684-132 77892-822
4,4'-(Propane-2,2-diyl)bis(2-isopropylphenol) ≥95%, Ambeed.Inc
4,4'-(Propane-2,2-diyl)bis(2-isopropylphenol)

Precaución: For research use only.

Formula: C₂₁H₂₈O₂
MDL Number: MFCD08276311
CAS Number: 127-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR