Tris(2-aminoethyl)amine ≥98%

Supplier: Ambeed
Danger

A397912-25G A397912-5G
77572-224EA 71 USD
77572-224 77872-624
Tris(2-aminoethyl)amine ≥98%
Tris(2-aminoethyl)amine

Precaución: for Research Use Only

Formula: C₆H₁₈N₄
MDL Number: MFCD00008177
CAS Number: 4097-89-6
UN: 2922
ADR: 8,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR