Boc-D-Dap(Z)-OH·DCHA ≥95%

Supplier: Ambeed
Warning

Synonyms: Dicyclohexylamine (R)-3-(((benzyloxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)propanoate

A397730-5G A397730-25G
77613-206EA 37.08 USD
77613-206 77686-103
Boc-D-Dap(Z)-OH·DCHA ≥95%
Boc-D-Dap(Z)-OH·DCHA

Precaución: for Research Use Only

Formula: C₂₈H₄₅N₃O₆
MW: 519.683 g/mol
MDL Number: MFCD00798629
CAS Number: 81306-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR