(S)-2-(N-Boc-aminomethyl)morpholine ≥95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl N-[(2S)-morpholin-2-ylmethyl]carbamate

A406722-100MG A406722-250MG
77872-642EA 26 USD
77872-642 77872-643
(S)-2-(N-Boc-aminomethyl)morpholine ≥95%
(S)-2-(N-Boc-aminomethyl)morpholine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₃
MW: 216.277 g/mol
MDL Number: MFCD12545849
CAS Number: 875551-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR