7-Bromo-2,3-dihydroquinolin-4(1H)-one ≥98%
Supplier: Ambeed
|
Warning
|
A406658-1G
A406658-250MG
77613-246EA
239.99
USD
77613-246
77684-147
7-Bromo-2,3-dihydroquinolin-4(1H)-one ≥98%
7-Bromo-2,3-dihydroquinolin-4(1H)-one
Precaución: For research use only.
Formula:
C₉H₈BrNO MW: 226.07 g/mol |
MDL Number:
MFCD12028725 CAS Number: 114417-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...