1H-Indole-6-carboxylic acid, 2,3-dihydro-2,3-dioxo-, methyl ester ≥98%

Supplier: Ambeed
Warning

A396878-1G A396878-250MG
77892-816EA 162 USD
77892-816 77892-817
1H-Indole-6-carboxylic acid, 2,3-dihydro-2,3-dioxo-, methyl ester ≥98%
1H-Indole-6-carboxylic acid, 2,3-dihydro-2,3-dioxo-, methyl ester

Precaución: for Research Use Only

Formula: C₁₀H₇NO₄
MDL Number: MFCD17214341
CAS Number: 213670-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR