Methyl 6-amino-2-methylnicotinate ≥95%

Supplier: Ambeed
Warning

A397147-100MG A397147-1G A397147-250MG A397147-5G
77886-055EA 50 USD
77886-055 77886-056 77886-057 77886-058
Methyl 6-amino-2-methylnicotinate ≥95%
Methyl 6-amino-2-methylnicotinate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD08062965
CAS Number: 872355-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR