L(+)-allo-Isoleucine ≥98%

Supplier: Ambeed
Warning

Synonyms: (2S,3R)-(+)-allo-Isoleucine, L-Alloisoleucine, (2S,3R)-2-Amino-3-methylpentanoic acid

A397030-10G A397030-1G A397030-250MG A397030-5G A397030-100MG
77686-096EA 928 USD
77686-096 77686-097 77686-098 77686-099 77892-818
L(+)-allo-Isoleucine ≥98%
L(+)-allo-Isoleucine

Precaución: for Research Use Only

Formula: H₃CCH₂CH(CH₃)CH(NH₂)COOH
MW: 131.17
MDL Number: MFCD00066446
CAS Number: 1509-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR