3-(Chloromethyl)-5-methylisoxazole ≥98%

Supplier: Ambeed
Danger

A396595-1G A396595-5G
77890-817EA 38 USD
77890-817 77890-818
3-(Chloromethyl)-5-methylisoxazole ≥98%
3-(Chloromethyl)-5-methylisoxazole

Precaución: for Research Use Only

Formula: C₅H₆ClNO
MDL Number: MFCD00221166
CAS Number: 35166-37-1
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR