tert-Butyl 9-amino-3-azaspiro[5.5]undecane-3-carboxylate ≥95%

Supplier: Ambeed
Warning

A387663-100MG A387663-250MG
77613-156EA 146.26 USD
77613-156 77613-158
tert-Butyl 9-amino-3-azaspiro[5.5]undecane-3-carboxylate ≥95%
tert-Butyl 9-amino-3-azaspiro[5.5]undecane-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₈N₂O₂
MW: 268.4 g/mol
MDL Number: MFCD18711593
CAS Number: 1272758-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR