Potassium 5-bromo-2-thiophenetrifluoroborate ≥98%

Supplier: Ambeed
Warning

Synonyms: Potassium (5-bromothiophen-2-yl)trifluoroborate

A396143-1G A396143-250MG A396143-5G
77901-932EA 55 USD
77901-932 77900-767 77901-153
Potassium 5-bromo-2-thiophenetrifluoroborate ≥98%
Potassium 5-bromo-2-thiophenetrifluoroborate

Precaución: for Research Use Only

Formula: C₄H₂BBrF₃S·K
MDL Number: MFCD17015269
CAS Number: 1239370-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR