EDTA tetrasodium salt dihydrate ≥99%

Supplier: Ambeed
Danger

Synonyms: Tetrasodium ethylenediaminetetraacetate dihydrate, Edetic acid tetrasodium salt dihydrate, Kestranal® 4S 2 H2O, Ethylenedinitrilotetra-acetic acid tetrasodium salt dihydrate, Ethylenediaminetetra-acetic acid tetrasodium salt dihydrate, Ethylenediaminetetraacetic acid sodium hydrate, Tetrasodium Ethylenediaminetetraacetate Dihydrate, Kestranal® 4S dihydrate

A387432-100G A387432-500G A387432-1000G
77601-706EA 10.25 USD
77601-706 77601-708 77872-606
EDTA tetrasodium salt dihydrate ≥99%
EDTA tetrasodium salt dihydrate

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂Na₄O₈·2H₂O
MW: 416.2 g/mol
MDL Number: MFCD00150026
CAS Number: 10378-23-1
Merck Index: 13,03544

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR