2,3,4-Trichloroaniline ≥98%

Supplier: Ambeed
Danger

A395771-1G A395771-5G
77572-214EA 26.78 USD
77572-214 77746-794
2,3,4-Trichloroaniline ≥98%
2,3,4-Trichloroaniline

Precaución: for Research Use Only

Formula: C₆H₄Cl₃N
MW: 196.46 g/mol
MDL Number: MFCD00042819
CAS Number: 634-67-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR