3,3'-Diamino-[1,1'-biphenyl]-4,4'-dicarboxylic acid ≥97%

Supplier: Ambeed
Warning

A386615-250MG A386615-1G
77748-779EA 33 USD
77748-779 77748-778
3,3'-Diamino-[1,1'-biphenyl]-4,4'-dicarboxylic acid ≥97%
3,3'-Diamino-[1,1'-biphenyl]-4,4'-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂O₄
MW: 272.25 g/mol
CAS Number: 1799740-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR