2-Chloro-4-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥96%, Ambeed.Inc

Supplier: Ambeed
Warning

A386384-1G A386384-100MG A386384-250MG A386384-5G
77684-112EA 157 USD
77684-112 77886-015 77886-016 77886-017
2-Chloro-4-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥96%, Ambeed.Inc
2-Chloro-4-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: For research use only.

Formula: C₁₁H₁₄BClFNO₂
MW: 257.5 g/mol
MDL Number: MFCD17926497
CAS Number: 1256359-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR