2,2-Dimethylcaproic acid ≥98%

Supplier: Ambeed
Danger

Synonyms: 2,2-Dimethylhexanoic acid

A395131-1G A395131-5G
77878-132EA 94 USD
77878-132 77878-133
2,2-Dimethylcaproic acid ≥98%
2,2-Dimethylcaproic acid

Precaución: for Research Use Only

Formula: C₈H₁₆O₂
MDL Number: MFCD00965097
CAS Number: 813-72-9
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR