Azithromycin dihydrate ≥99%

Supplier: Ambeed
Warning

Synonyms: Azithromycin hydrate, (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-(((2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-2-ethyl-3,4,10-trihydroxy-13-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-3,5,6,8,10,12,14-heptamethyl-1-oxa-6-azacyclopentadecan-15-one dihydrate

A385826-10MG A385826-1MG A385826-50MG
77626-764EA 6.18 USD
77626-764 77626-766 77626-768
Azithromycin dihydrate ≥99%
Azithromycin dihydrate

Precaución: for Research Use Only

Formula: C₃₈H₇₂N₂O₁₂·2H₂O
MW: 749.00 g/mol
MDL Number: MFCD01862248
CAS Number: 117772-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR