Phenanthrene-9-carboxylic acid ≥97%

Supplier: Ambeed
Warning

A394587-250MG A394587-1G
77601-740EA 27.68 USD
77601-740 77668-824
Phenanthrene-9-carboxylic acid ≥97%
Phenanthrene-9-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₀O₂
MDL Number: MFCD00019111
CAS Number: 837-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR