cis-4-Heptenal ≥98%

Supplier: Ambeed
Danger

Synonyms: cis-4-Heptenal, Cis-4-heptenal, (Z)-4-Heptenal

A394581-1G A394581-25G A394581-5G
77601-736EA 26.65 USD
77601-736 77601-738 77626-806
cis-4-Heptenal ≥98%
cis-4-Heptenal

Precaución: for Research Use Only

Formula: C₇H₁₂O
MW: 112.17 g/mol
MDL Number: MFCD00014682
CAS Number: 6728-31-0
UN: 1989
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR