3-Bromopyridine-2-thiol ≥98%

Supplier: Ambeed
Warning

A385408-1G A385408-250MG
77892-789EA 78 USD
77892-789 77892-790
3-Bromopyridine-2-thiol ≥98%
3-Bromopyridine-2-thiol

Precaución: for Research Use Only

Formula: C₅H₄BrNS
MDL Number: MFCD14651478
CAS Number: 65938-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR