(9R,10R)-9-Acetoxy-8,8-dimethyl-2-oxo-9,10-dihydro-2H,8H-pyrano[2,3-f]chromen-10-yl (Z)-2-methylbut-2-enoate ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pteryxin
A394213-1MG
A394213-10MG
A394213-5MG
77613-196EA
10.3
USD
77613-196
77684-128
77684-129
(9R,10R)-9-Acetoxy-8,8-dimethyl-2-oxo-9,10-dihydro-2H,8H-pyrano[2,3-f]chromen-10-yl (Z)-2-methylbut-2-enoate ≥98%
(9R,10R)-9-Acetoxy-8,8-dimethyl-2-oxo-9,10-dihydro-2H,8H-pyrano[2,3-f]chromen-10-yl (Z)-2-methylbut-2-enoate
Precaución: For research use only.
Formula:
C₂₁H₂₂O₇ MW: 386.4 g/mol |
MDL Number:
MFCD11519465 CAS Number: 13161-75-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...