5'-O-DMT-2'-TBDMS-Uridine ≥97%

Supplier: Ambeed
Warning

Synonyms: 1-((2R,3R,4R,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-3-((tert-butyldimethylsilyl)oxy)-4-hydroxytetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione

A384927-5G A384927-1G A384927-250MG
77572-144EA 219.39 USD
77572-144 77594-162 77613-148
5'-O-DMT-2'-TBDMS-Uridine ≥97%
5'-O-DMT-2'-TBDMS-Uridine

Precaución: for Research Use Only

Formula: C₃₆H₄₄N₂O₈Si
MW: 660.83 g/mol
MDL Number: MFCD00135726
CAS Number: 81246-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR