(1R,2R)-2-aminocyclobutan-1-ol hydrochloride ≥97%

Supplier: Ambeed
Warning

A393817-250MG A393817-1G
77748-790EA 570 USD
77748-790 77748-789
(1R,2R)-2-aminocyclobutan-1-ol hydrochloride ≥97%
(1R,2R)-2-aminocyclobutan-1-ol hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClNO
MW: 123.58 g/mol
CAS Number: 1909287-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR