Di-tert-butyl pyrazolidine-1,2-dicarboxylate 95+%
Supplier: Ambeed
|
Warning
|
A393368-1G
A393368-5G
77900-494EA
76
USD
77900-494
77901-931
Di-tert-butyl pyrazolidine-1,2-dicarboxylate 95+%
Di-tert-butyl pyrazolidine-1,2-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₃H₂₄N₂O₄ |
MDL Number:
MFCD16620914 CAS Number: 146605-64-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...